MAT238926309 Matrix Scientific

(4-Bromophenyl)(cyclopropyl)ketone

  • MDL number MFCD00019227
  • Cas Number : 6952-89-2
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043959.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043959.html
Availability details for
Quantity
  • Availability
  • Add to Cart