MAT238926342 Matrix Scientific

1-(3-Ethynylphenyl)-3,3-dimethyl-2-azetanone

  • MDL number MFCD06200947
  • Cas Number : 886361-64-4
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-59
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043992.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043992.html
Availability details for
Quantity
  • Availability
  • Add to Cart