MAT238926384 Matrix Scientific

4-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]benzenol

  • MDL number MFCD00138769
  • Cas Number : 75998-99-1
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232-234
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/044034.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/044034.html
Availability details for
Quantity
  • Availability
  • Add to Cart