MAT238926395 Matrix Scientific

5-(Carboxymethyl)-1-(4-chlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD05021854
  • Cas Number : 439095-10-0
  • Molecular Weight : 281.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/044045.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/044045.html
Availability details for
Quantity
  • Availability
  • Add to Cart