MAT239351862 Matrix Scientific

Ethyl 5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD03407995
  • Cas Number : 155377-19-8
  • Molecular Weight : 208.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-148
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/006377.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006377.html
Availability details for
Quantity
  • Availability
  • Add to Cart