MAT239355244 Matrix Scientific

4-Chloro-2-pyridinecarbaldehyde

  • MDL number MFCD07437896
  • Cas Number : 63071-13-6
  • Molecular Weight : 141.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-34
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/048191.html

Safety Information

hcodes Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart