MAT240053255 Matrix Scientific

4-Azepan-1-ylaniline dihydrochloride

  • MDL number MFCD09431467
  • Cas Number : 57356-18-0
  • Molecular Weight : 263.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/030933.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/030933.html
Availability details for
Quantity
  • Availability
  • Add to Cart