MAT242635081 Matrix Scientific

4-Methyl-2-propoxyaniline hydrochloride

  • MDL number MFCD13193065
  • Cas Number : 640767-75-5
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022186.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022186.html
Availability details for
Quantity
  • Availability
  • Add to Cart