MAT246514017 Matrix Scientific

Tricyclo[4.3.1.1~3,8~]undec-3-ylaminehydrochloride

  • MDL number MFCD00267989
  • Cas Number : 3048-63-3
  • Molecular Weight : 201.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/038588.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/038588.html
Availability details for
Quantity
  • Availability
  • Add to Cart