MAT246517634 Matrix Scientific

N~1~,N~1~-dimethyl-1,2-benzenediamine

  • MDL number MFCD01706706
  • Cas Number : 2836-03-5
  • Molecular Weight : 136.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 218
Product Link http://www.matrixscientific.com/051072.html

Safety Information

hcodes Irritant/Light Sensitive/Store in Freezer
MSDS http://www.matrixscientific.com/051072.html
Availability details for
Quantity
  • Availability
  • Add to Cart