MAT267321888 Matrix Scientific

1,2,3,4-Tetrahydro-isoquinoline-1-carboxylic acidethyl ester hydrochloride

  • MDL number MFCD08689587
  • Cas Number : 103733-33-1
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-137
Country of Origin USA
Product Link http://www.matrixscientific.com/008250.html

Safety Information

MSDS http://www.matrixscientific.com/008250.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart