MAT293039539 Matrix Scientific

4-Amino-N-(2-aminoethyl)-1,2,5-oxadiazole-3-carboxamide hydrochloride

  • MDL number MFCD16613946
  • Cas Number : 1228070-89-0
  • Molecular Weight : 207.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/011503.html

Safety Information

MSDS http://www.matrixscientific.com/011503.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart