MAT309666408 Matrix Scientific

2-(4-Amino-1H-pyrazol-1-yl)acetohydrazidehydrochloride

  • MDL number MFCD17676097
  • Cas Number : 1256264-87-5
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/040754.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/040754.html
Availability details for
Quantity
  • Availability
  • Add to Cart