MAT309666754 Matrix Scientific

1-Benzyl-2-thiourea

  • MDL number MFCD00041370
  • Cas Number : 621-83-0
  • Molecular Weight : 166.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-166
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/056653.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/056653.html
Availability details for
Quantity
  • Availability
  • Add to Cart