MAT309667420 Matrix Scientific

N-Allyl-N-[2-(allyloxy)phenyl]propanamide

  • MDL number MFCD17676106
  • Cas Number : 1246471-49-7
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058418.html

Safety Information

MSDS http://www.matrixscientific.com/058418.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart