MAT309668373 Matrix Scientific

3-(Benzyloxy)-N-(sec-butyl)benzamide

  • MDL number MFCD08593289
  • Cas Number : 908551-06-4
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058416.html

Safety Information

MSDS http://www.matrixscientific.com/058416.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart