MAT309668400 Matrix Scientific

6-Cyclohexyl-2,3,4,9-tetrahydro-1H-carbazol-1-one

  • MDL number MFCD00427056
  • Cas Number : 135897-70-0
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058444.html

Safety Information

MSDS http://www.matrixscientific.com/058444.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart