MAT309668408 Matrix Scientific

5-(Benzyloxy)tryptophan

  • MDL number MFCD00037968
  • Cas Number : 1956-25-8
  • Molecular Weight : 310.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 275-276
Country of Origin USA
Product Link http://www.matrixscientific.com/058453.html

Safety Information

MSDS http://www.matrixscientific.com/058453.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart