MAT309668422 Matrix Scientific

1,2,3,4-Naphthalenetetrone

  • MDL number MFCD00985549
  • Cas Number : 30266-58-1
  • Molecular Weight : 188.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058468.html

Safety Information

MSDS http://www.matrixscientific.com/058468.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart