MAT309668438 Matrix Scientific

5-[(4-Chlorophenyl)methyl]-1,3,4-oxadiazol-2-ol

  • MDL number MFCD17392925
  • Cas Number : 34547-04-1
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058493.html

Safety Information

MSDS http://www.matrixscientific.com/058493.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart