MAT309668440 Matrix Scientific

3-Hydrazono-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD00170809
  • Cas Number : 2365-44-8
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058457.html

Safety Information

MSDS http://www.matrixscientific.com/058457.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart