MAT311774052 Matrix Scientific

1-Fluoro-4-nitro-2-(2,2,2-trifluoroethyl)-benzene

  • MDL number MFCD18384827
  • Cas Number : 1262413-56-8
  • Molecular Weight : 223.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058593.html

Safety Information

MSDS http://www.matrixscientific.com/058593.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart