MAT311774085 Matrix Scientific

4-(4-Trifluoromethyl-pyridin-2-yl)-benzoic acid

  • MDL number MFCD18384864
  • Cas Number : 1299607-71-8
  • Molecular Weight : 267.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059272.html

Safety Information

MSDS http://www.matrixscientific.com/059272.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart