MAT311774092 Matrix Scientific

2-Butenamide

  • MDL number MFCD00059859
  • Cas Number : 23350-58-5
  • Molecular Weight : 85.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/058466.html

Safety Information

MSDS http://www.matrixscientific.com/058466.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart