MAT311774107 Matrix Scientific

3-Piperazin-1-yl-1,2-benzisoxazole

  • MDL number MFCD08060968
  • Cas Number : 87691-89-2
  • Molecular Weight : 203.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058606.html

Safety Information

MSDS http://www.matrixscientific.com/058606.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart