MAT311774146 Matrix Scientific

1,3-Dimethyl-1H,4H,5H,6H,7H-pyrazolo[3,4-d]-pyrimidine-4,6-dione

  • MDL number MFCD18157655
  • Cas Number : 1072895-79-4
  • Molecular Weight : 180.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058645.html

Safety Information

MSDS http://www.matrixscientific.com/058645.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart