MAT311774147 Matrix Scientific

Ethyl-2-chloro-2-[2-(4-methoxyphenyl)hydrazin-1-ylidene]acetate

  • MDL number MFCD00446053
  • Cas Number : 27143-07-3
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058646.html

Safety Information

MSDS http://www.matrixscientific.com/058646.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart