MAT311774316 Matrix Scientific

6-Bromo-2-trifluoromethyl-1H-benzoimidazole

  • MDL number MFCD00460102
  • Cas Number : 3671-60-1
  • Molecular Weight : 265.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058815.html

Safety Information

MSDS http://www.matrixscientific.com/058815.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart