MAT311774324 Matrix Scientific

1-tert-Butyl-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD01313628
  • Cas Number : 376387-68-7
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058823.html

Safety Information

MSDS http://www.matrixscientific.com/058823.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart