MAT311774342 Matrix Scientific

3-Chloro-2-chloromethyl-5-(trifluoromethyl)pyridine

  • MDL number MFCD00173852
  • Cas Number : 175277-52-8
  • Molecular Weight : 230.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/058841.html

Safety Information

MSDS http://www.matrixscientific.com/058841.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart