MAT311774367 Matrix Scientific

4-Chloro-3-methyl-isothiazol-5-ylamine

  • MDL number MFCD09909657
  • Cas Number : 96841-04-2
  • Molecular Weight : 148.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058866.html

Safety Information

MSDS http://www.matrixscientific.com/058866.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart