MAT311774368 Matrix Scientific

4-Chloro-5-methyl-isoxazol-3-ylamine

  • MDL number MFCD11227222
  • Cas Number : 5819-39-6
  • Molecular Weight : 132.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058867.html

Safety Information

MSDS http://www.matrixscientific.com/058867.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart