MAT311774379 Matrix Scientific

2-Chloro-5-methyl-pyridine-3-sulfonyl chloride

  • MDL number MFCD15142818
  • Cas Number : 1208081-98-4
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058878.html

Safety Information

MSDS http://www.matrixscientific.com/058878.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart