MAT311774447 Matrix Scientific

4-(3-Cyano-pyridin-2-yl)-benzoic acid ethyl ester

  • MDL number MFCD15142822
  • Cas Number : 1208081-99-5
  • Molecular Weight : 252.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/058946.html

Safety Information

MSDS http://www.matrixscientific.com/058946.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart