MAT311774529 Matrix Scientific

Ethyl 1-carbamimidoylpiperidine-4-carboxyateacetate

  • MDL number MFCD10568351
  • Cas Number : 1208081-80-4
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059028.html

Safety Information

MSDS http://www.matrixscientific.com/059028.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart