MAT311774623 Matrix Scientific

1-(4-Methoxy-pyridin-2-yl)-ethanone

  • MDL number MFCD10697664
  • Cas Number : 59576-28-2
  • Molecular Weight : 151.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059122.html

Safety Information

MSDS http://www.matrixscientific.com/059122.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart