MAT311774708 Matrix Scientific

3-Oxo-pyrrolidine-1,2-dicarboxylic acid1-tert-butyl ester 2-ethyl ester

  • MDL number MFCD11052440
  • Cas Number : 170123-25-8
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059207.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/059207.html
Availability details for
Quantity
  • Availability
  • Add to Cart