MAT311774722 Matrix Scientific

6-Phenyl-pyrimidine-2,4-diol

  • MDL number MFCD00128171
  • Cas Number : 13345-09-0
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059221.html

Safety Information

MSDS http://www.matrixscientific.com/059221.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart