MAT311774766 Matrix Scientific

5-(Trifluoromethyl)pyridine-2-thioamide

  • MDL number MFCD00205810
  • Cas Number : 175277-51-7
  • Molecular Weight : 206.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059265.html

Safety Information

MSDS http://www.matrixscientific.com/059265.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart