MAT316263483 Matrix Scientific

1-Ethyl-2-methyl-1H-benzoimidazol-5-ylaminehydrochloride

  • MDL number MFCD03939015
  • Cas Number : 1172850-46-2
  • Molecular Weight : 211.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020402.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020402.html
Availability details for
Quantity
  • Availability
  • Add to Cart