MAT321702301 Matrix Scientific

Methyl (2S,4S)-4-(2-chloro-5-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

  • MDL number MFCD18803478
  • Cas Number : 1354487-97-0
  • Molecular Weight : 310.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023705.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023705.html
Availability details for
Quantity
  • Availability
  • Add to Cart