MAT321702305 Matrix Scientific

Methyl 6,7-dimethoxy-1,2,3,4-tetra-hydroisoquinoline-3-carboxylate hydrochloride

  • MDL number MFCD08594063
  • Cas Number : 30740-96-6
  • Molecular Weight : 287.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031044.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031044.html
Availability details for
Quantity
  • Availability
  • Add to Cart