MAT321702311 Matrix Scientific

3-Fluoro-5-methoxypyridine

  • MDL number MFCD13185606
  • Cas Number : 1060801-62-8
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/059326.html

Safety Information

MSDS http://www.matrixscientific.com/059326.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart