MAT321702564 Matrix Scientific

6-Ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD18377697
  • Cas Number : 15043-03-5
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-209
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/059579.html

Safety Information

MSDS http://www.matrixscientific.com/059579.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart