MAT321761308 Matrix Scientific

2-Ethyl-6-methyl-3-pyridinyl dimethylcarbamatesuccinate (2:1)

  • MDL number MFCD18800410
  • Cas Number : 205189-71-5
  • Molecular Weight : 534.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Country of Origin USA
Product Link http://www.matrixscientific.com/059743.html

Safety Information

MSDS http://www.matrixscientific.com/059743.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart