MAT321761316 Matrix Scientific

Sodium 1-hydroxy-2-oxo-2-(9-oxo-9H-fluoren-2-yl)-ethanesulfonate

  • MDL number MFCD01970967
  • Cas Number : 51367-77-2
  • Molecular Weight : 340.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059678.html

Safety Information

MSDS http://www.matrixscientific.com/059678.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart