MAT321761319 Matrix Scientific

Methyl (4-{[(5-ethyl-1,3,4-thiadiazol-2-yl)amino]-sulfonyl}phenyl)carbamate

  • MDL number MFCD00475787
  • Cas Number : 35746-80-6
  • Molecular Weight : 342.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059681.html

Safety Information

MSDS http://www.matrixscientific.com/059681.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart