MAT321761325 Matrix Scientific

3,12-Bis(2-chloroethyl)-3,12-diaza-6,9-diazoniadispiro[5.2.5.2]hexadecane dichloride

  • MDL number MFCD18909629
  • Cas Number : 1158-80-1
  • Molecular Weight : 422.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059687.html

Safety Information

MSDS http://www.matrixscientific.com/059687.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart