MAT321761326 Matrix Scientific

N,N-Dimethyl-2-(10H-phenothiazin-10-yl)ethanaminehydrochloride

  • MDL number MFCD01706893
  • Cas Number : 5934-20-3
  • Molecular Weight : 306.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059688.html

Safety Information

MSDS http://www.matrixscientific.com/059688.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart