MAT321761335 Matrix Scientific

2-(1-Cyclohexyl-2-piperazinyl)ethanol

  • MDL number MFCD16885711
  • Cas Number : 1211505-12-2
  • Molecular Weight : 212.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/059697.html

Safety Information

MSDS http://www.matrixscientific.com/059697.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart